missing translation for 'onlineSavingsMsg'
Learn More

FGF-21 Antibody (461801) [DyLight 594], Novus Biologicals™

Product Code. 30736783 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736783 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736783 Supplier Novus Biologicals Supplier No. FAB25374M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-21
Applications ELISA
Classification Monoclonal
Clone 461801
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FGF-21, fibroblast growth factor 21
Host Species Mouse
Immunogen E. coli-derived recombinant human FGF-21, His29-Ser209, Accession # Q9NSA1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 26291
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.