missing translation for 'onlineSavingsMsg'
Learn More

FGF-20 Antibody (272317) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30734076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734076 Supplier Novus Biologicals Supplier No. FAB2547CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

FGF-20 Monoclonal antibody specifically detects FGF-20 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-20
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 272317
Conjugate CoraFluor 1
Formulation PBS
Gene Alias FGF-20, fibroblast growth factor 20
Host Species Rat
Immunogen E. coli-derived recombinant human FGF-20, Met1-Thr211, Accession # Q9NP95
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26281
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.