missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-19 Antibody (MM0285-4G16) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£153.00 - £423.00
Specifications
| Antigen | FGF-19 |
|---|---|
| Clone | MM0285-4G16 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18058766
|
Novus Biologicals
NBP2-12299-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18751604
|
Novus Biologicals
NBP2-12299 |
0.1 mg | |||||||
Description
FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FGF-19 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FGF-19, fibroblast growth factor 19 | |
| FGF19 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| MM0285-4G16 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Neuroscience | |
| O95750 | |
| 9965 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title