missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress FGF-16 Protein, Human (CHO)
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Content And Storage | Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage. |
| Quantity | 10 μg |
| Shipping Condition | Room temperature |
| Molecular Weight (g/mol) | Approximately 23 kDa |
| Synonym | rHuFGF-16∣ FGF-16∣ Fibroblast Growth Factor-16 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction