Learn More
FGF-16 Antibody (MM0283-6G2), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-12297PCP
Product Code. 18462322
  • £460.00 / 0.10mL
Description
FGF-16 Monoclonal antibody specifically detects FGF-16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF-16
Monoclonal
PerCP
PBS
Mouse
Protein G purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0283-6G2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FGF-16, fibroblast growth factor 16
Human recombinant FGF-16
0.1 mL
Angiogenesis
8823
Store at 4C in the dark.
IgG2