missing translation for 'onlineSavingsMsg'
Learn More

FGF-15 Antibody [FITC], Novus Biologicals™

Product Code. 30736295 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736295 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736295 Supplier Novus Biologicals Supplier No. AF6755F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

FGF-15 Polyclonal antibody specifically detects FGF-15 in Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-15
Applications Immunohistochemistry
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias FGF-19
Host Species Sheep
Immunogen E. coli-derived recombinant mouse FGF-15, Ser24-Lys218, Accession # O35622
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9965
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.