missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-13 Antibody (S235-22) - Pan Specific, Novus Biologicals™
Mouse Monoclonal Antibody
£168.00 - £427.00
Specifications
| Antigen | FGF-13 |
|---|---|
| Clone | S235-22 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610957
|
Novus Biologicals
NBP2-42178 |
0.1 mg |
£427.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635847
|
Novus Biologicals
NBP2-42178-0.025mg |
0.025 mg |
£168.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-13 Monoclonal antibody specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| FGF-13 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Biologically Active Proteins | |
| PBS (pH 7.4), 50% Glycerol | |
| FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
| Synthetic peptide amino acids 2-18 (AAAIASSLIRQKRQARE) of human FHF2A. 100% identical to rat, 94% identical to mouse. >80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S235-22 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AAH34340 | |
| 2258 | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title