missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-13 Antibody (OTI7E8), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70721V
This item is not returnable.
View return policy
Description
FGF-13 Monoclonal specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FGF-13 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FGF13 | |
| Full length recombinant protein of human FGF-13 (NP_004105) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7E8 | |
| Western Blot, Immunohistochemistry | |
| FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Biologically Active Proteins, Neuroscience | |
| 2258.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction