missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-13 Antibody (298923) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1909JF525
This item is not returnable.
View return policy
Description
FGF-13 Monoclonal antibody specifically detects FGF-13 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| FGF-13 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
| E. coli-derived recombinant human FGF-13 isoform 1B, Ala2-Thr192, Accession # NP_378668 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Neuroscience | |
| 2258 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Western Blot, CyTOF | |
| 298923 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction