missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-13 1B Antibody, R&D Systems™
Goat Polyclonal Antibody
£418.32
Specifications
Antigen | FGF-13 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FGF-13 Polyclonal specifically detects FGF-13 in Human samples. It is validated for Western Blot.Specifications
FGF-13 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
FGF13, FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
FGF13 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q92913-2 | |
2258 | |
E. coli-derived recombinant human FGF-13 isoform 1B Ala2-Thr192 Accession # Q92913-2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human FGF-13 1B in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) FGF-12 is observed and 5% cross-reactivity with rhFGF-11 is observed. |
For Research Use Only