missing translation for 'onlineSavingsMsg'
Learn More

FGF-12 Antibody (1D9), Novus Biologicals™

Product Code. 18383798 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383798 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383798 Supplier Novus Biologicals Supplier No. H00002257M10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-12 Monoclonal antibody specifically detects FGF-12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-12
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1D9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH22524
Gene Alias FGF-12, FGF12B, FHF-1, FHF1Fibroblast growth factor homologous factor 1, fibroblast growth factor 12, fibroblast growth factor 12B, fibroblast growth factor FGF-12b, Myocyte-activating factor
Host Species Mouse
Immunogen FGF12 (AAH22524, 1 a.a. ~ 181 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MESKEPQLKGIVTRLFSQQGYFLQMHPDGTIDGTKDENSDYTLFNLIPVGLRVVAIQGVKASLYVAMNGEGYLYSSDVFTPECKFKESVFENYYVIYSSTLYRQQESGRAWFLGLNKEGQIMKGNRVKKTKPSSHFVPKPIEVCMYREQSLHEIGEKQGRSRKSSGTPTMNGGKVVNQDST
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 2257
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.