missing translation for 'onlineSavingsMsg'
Learn More

FGF-11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£285.00 - £423.00

Specifications

Antigen FGF-11
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18266921
View Documents
Novus Biologicals
NBP2-55935
100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18680979
View Documents
Novus Biologicals
NBP2-55935-25ul
25 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

FGF-11 Polyclonal specifically detects FGF-11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

FGF-11
Polyclonal
Rabbit
Angiogenesis, Neuroscience
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
2256
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VTKLFCRQGFYLQANPDGSIQGTPEDTSSFTHFN
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
FGF-11, FHF-3, FHF3Fibroblast growth factor homologous factor 3, fibroblast growth factor 11, FLJ16061, MGC102953, MGC45269
FGF11
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.