missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-11 Antibody (MM0282-6J20), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12296PCP
This item is not returnable.
View return policy
Description
FGF-11 Monoclonal antibody specifically detects FGF-11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FGF-11 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0282-6J20 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FGF-11, FHF-3, FHF3Fibroblast growth factor homologous factor 3, fibroblast growth factor 11, FLJ16061, MGC102953, MGC45269 | |
| Human recombinant FGF-11 | |
| 0.1 mL | |
| Angiogenesis, Neuroscience | |
| 2256 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction