missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FGD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29956
This item is not returnable.
View return policy
Description
FGD3 Polyclonal antibody specifically detects FGD3 in Human samples. It is validated for Immunoprecipitation
Specifications
| FGD3 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 89846 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| FGD1 family, member 3, FLJ00004, FYVE, RhoGEF and PH domain containing 3, FYVE, RhoGEF and PH domain-containing protein 3, GCCD2, GCCD3, MGC117260, ZFYVE5faciogenital dysplasia 3, Zinc finger FYVE domain-containing protein 5 | |
| Between 50 and 100 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction