missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585408
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (98%), Bovine (100%). Positive Control: Target recombinant protein, U87-MG, SK-N-SH, IMR32, SK-N-AS. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FGD1 contains Dbl (DH) and pleckstrin (PH) homology domains. It can bind specifically to the Rho family GTPase Cdc42Hs and stimulate the GDP-GTP exchange of the isoprenylated form of Cdc42Hs. It also stimulates the mitogen activated protein kinase cascade leading to c-Jun kinase SAPK/JNK1 activation. FGD1 has an essential role in embryonic development, and FGD1 gene mutations result in the human developmental disorder, Aarskog-Scott syndrome.
Specifications
| FGD1 | |
| Polyclonal | |
| Unconjugated | |
| FGD1 | |
| AAS; faciogenital dysplasia 1 protein; Faciogenital dysplasia 1 protein homolog; faciogenital dysplasia protein; Fgd1; FGDY; FYVE, RhoGEF and PH domain containing 1; FYVE, RhoGEF and PH domain containing 1 (faciogenital dysplasia); FYVE, RhoGEF and PH domain-containing protein 1; MRXS16; Rho/Rac GEF; rho/Rac guanine nucleotide exchange factor FGD1; ZFYVE3; zinc finger FYVE domain-containing protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2245 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.85 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P98174 | |
| FGD1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human FGD1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction