missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541477
This item is not returnable.
View return policy
Description
Peptide sequence: HGHRAPGGAG PSEPEHPATN PPGAAPPACA DSDPGASEPG LLARRGSGSA Sequence homology: Cow: 86%; Human: 100%; Pig: 86%; Rat: 79%.
FGD1 contains Dbl (DH) and pleckstrin (PH) homology domains. It can bind specifically to the Rho family GTPase Cdc42Hs and stimulate the GDP-GTP exchange of the isoprenylated form of Cdc42Hs. It also stimulates the mitogen activated protein kinase cascade leading to c-Jun kinase SAPK/JNK1 activation. FGD1 has an essential role in embryonic development, and FGD1 gene mutations result in the human developmental disorder, Aarskog-Scott syndrome.
Specifications
| FGD1 | |
| Polyclonal | |
| Unconjugated | |
| FGD1 | |
| AAS; faciogenital dysplasia 1 protein; Faciogenital dysplasia 1 protein homolog; faciogenital dysplasia protein; Fgd1; FGDY; FYVE, RhoGEF and PH domain containing 1; FYVE, RhoGEF and PH domain containing 1 (faciogenital dysplasia); FYVE, RhoGEF and PH domain-containing protein 1; MRXS16; Rho/Rac GEF; rho/Rac guanine nucleotide exchange factor FGD1; ZFYVE3; zinc finger FYVE domain-containing protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2245 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P98174 | |
| FGD1 | |
| Synthetic peptide directed towards the N-terminal of human FGD1 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction