Learn More
Chemsplash FFP3 Fold Flat Mask with Valve
GREENER_CHOICE
Product Code. 30058357
Change view
Click to view available options
Aerosol Type:
R - Somewhat Resistant to Oil
Unit Size:
Pack of 200
Product Code. Aerosol Type Unit Size
30058357 R - Somewhat Resistant to Oil Pack of 200
1 options
Product Code. 30058357 Supplier Chemsplash Supplier No. 3675
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
The FPP3 valved fold flat mask has a high filtering efficiency. It has a woven elasticated adjustable toggles.
Chemsplash 3675 P3 Fold Flat Mask
  • High Filtration Capacity
  • Respirator with Red Color Exhalation Valve
  • Exhalation Valve Reduces Hot Air Buildup
  • Woven Elasticated Adjustable Head Straps
  • Adjustable Nose Clip with Soft Inner Nose Foam
  • Folded to Pocket Size and Packed Individually for Hygiene
  • Protects Against Harmful and Toxic Solid and Liquid Particles
  • No Vision Impairment and Problem-Free Speech
  • Latex and Graphene Free
Description: The Chemsplash 3675 Fold Flat Mask is a P3 valved fold-flat respirator, color-coded red for easy identification. This mask offers a high filtration capacity and features woven elasticated adjustable toggles for a comfortable fit. It includes an adjustable nose clip with soft inner foam to ensure an excellent seal, minimizing air leakage. The mask is folded to pocket size and packed individually for hygiene. It effectively protects against harmful and toxic solid and liquid particles, ensuring no vision impairment or speech difficulty. Additionally, it is latex and graphene-free for safer use.
Specifications
Aerosol Type R - Somewhat Resistant to Oil
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile