Learn More
Chemsplash FFP3 Cup Mask with Valve
GREENER_CHOICE
Product Code. 30058326
Change view
Click to view available options
Aerosol Type:
R - Somewhat Resistant to Oil
Unit Size:
Pack of 60
Product Code. Aerosol Type unitSize
30058326 R - Somewhat Resistant to Oil Pack of 60
1 options
Product Code. 30058326 Supplier Chemsplash Supplier No. 3492
Only null left
Add to basket
Add to basket
The Valved Cup Dust Mask is colour coded red for ease of use. The fabric material of the mask has a high filtering efficiency.
Chemsplash 3492 Valved Cup Dust Mask
  • High Filtration Capacity
  • Red Color Exhalation Valve
  • Exhalation Valve Reduces Hot Air Buildup
  • Full Inner Face Seal with Excellent Fit
  • Plastic Welded Woven Elasticated Adjustable Head Straps
  • Protects Against Harmful and Toxic Solid and Liquid Particles
  • No Vision Impairment and Problem Free Speech
  • Latex and Graphene Free
Description: The Chemsplash 3492 is a Valved Cup Dust Mask, colour coded red for ease of use. The fabric material of the mask has high filtering efficiency, and it features plastic welded woven elasticated adjustable head straps for a secure fit. It also provides a full inner face seal for an excellent fit and helps reduce hot air buildup with the red exhalation valve. The mask protects against harmful and toxic solid and liquid particles, and it does not impair vision or speech. Latex and graphene-free, it is suitable for various industrial and hazardous environments.
Specifications
Aerosol Type R - Somewhat Resistant to Oil
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile