Learn More
Sperian™ Honeywell™ FFP2 ML Respirator Half Mask with Valve
Product Code. 11821781
Change view
Click to view available options
Model:
FFP2D
Unit Size:
Pack of 10
Product Code. Model unitSize
11821781 FFP2D Pack of 10
1 options
Product Code. 11821781 Supplier Sperian™ Supplier No. 1005614
Only null left
Add to basket
Edge
Add to basket
Protection and wearer acceptance guaranteed during periods of extended use. Sperian™ Honeywell™ FFP2 ML Respirator Half Mask with Valve provides an exceptional level of comfort and prevents irritation due to no latex allergy risk.
  • Folding respirator packed in a tightly sealed individual bag to prevent contamination
  • Fluid-resistant outer shell to reduce risk of contamination
  • Ultra-soft hypoallergenic Willtech™ nose seal with absorption properties and a tight sealing film
  • Preformed and colored nose bridge to save time to perfectly adjust the mask and identify its protection level
  • Large internal volume with built-in chin piece to easily communicate and breathe
  • Shielded exhalation valve with an exclusive design for improved protection and better breathing comfort
  • Elasticated sliding straps, highly flexible and resistant, to increase safety, comfort and ease of donning/doffing
  • No latex, no silicone, no PVC
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Product Type Filtering Half Mask for Single Use
Size Medium
With Exhalation Valve Yes
For Use With (Application) Textile, Mining, Construction, Iron and Steel industries, Car Body Preparation Industry, Woodworking.
Latex Free Yes
Material (Faceshield) PES knit, PU foam, PU film
Model FFP2D
Nose Piece Material Polypropylene-wrapped Galvanized wire
Quantity 10/Pk.
Resistant To Fluid
With Faceseal Yes
Weight (Metric) 14 g