missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ 5000 Series Specialty Disposable Respirators, FFP1-FFP2
Description
- Product Type: Specialty Respirator
- Disposable: Yes
- Description: Half-Mask
- Shape: Moulded
- Latex-Free: Yes
- Silicone-Free: Yes
- PVC-Free: Yes
Specifications
Specifications
| Certifications/Compliance (Europe) | EC Category: 3, EN 149:2001+A1:2009 |
| Color | Blue Stripe |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Product Type | Particulate Respirator with Nuisance Organic Vapor Relief |
| Size | Medium/Large |
| With Exhalation Valve | No |
| Filter Efficiency | FFP2 |
| For Use With (Application) | Textile, Mining, Abattoirs, Construction, Iron and Steel Industries, Car Body Preparation Industry, Woodworking |
| Latex Free | Yes |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?