missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ 5000 Series Specialty Disposable Respirators, FFP1-FFP2

Product Code. 11807010
Change view
Click to view available options
Product Type:
Particulate Respirator with Nuisance Acid Gas Relief
Particulate Respirator with Nuisance Organic Vapor Relief
Filter Efficiency:
FFP1
FFP2
Size:
Medium/Large
X-Large
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 20
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Filter Efficiency Size With Exhalation Valve unitSize
11807010 Particulate Respirator with Nuisance Acid Gas Relief FFP1 Medium/Large Yes Pack of 20
11836960 Particulate Respirator with Nuisance Organic Vapor Relief FFP1 Medium/Large Yes Pack of 20
11897000 Particulate Respirator with Nuisance Organic Vapor Relief FFP2 Medium/Large No Pack of 5
15165173 Particulate Respirator with Nuisance Organic Vapor Relief FFP2 X-Large No Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11807010 Supplier Honeywell™ Supplier No. 1005598
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

5000 Series Specialty Disposable Respirators, FFP1-FFP2 are moulded half-masks designed specifically to deal with nuisance level organic vapors and acid vapors. Latex-, silicone-, and PVC-free, they offer superior odor removal, high moisture absorption, and excellent comfort. Dolomite tested.

  • Product Type: Specialty Respirator
  • Disposable: Yes
  • Description: Half-Mask
  • Shape: Moulded
  • Latex-Free: Yes
  • Silicone-Free: Yes
  • PVC-Free: Yes
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EC Category: 3, EN 149:2001+A1:2009
Color Brown Stripe
Design Type Cup/Molded
Disposable Single-use
Product Type Particulate Respirator with Nuisance Acid Gas Relief
Size Medium/Large
With Exhalation Valve Yes
Filter Efficiency FFP1
Latex Free Yes
Material (Faceshield) PES Knit, PU Foam, PU Film
Model 5161
Nose Piece Material Polypropylene-wrapped Galvanized Wire
Nuisance Odor Relief Yes
Quantity 20 Pk.
Resistant To Fluid
Sterility Non-sterile
Style Specialty Half-Mask Respirator
With Faceseal Yes (Nasal Seal)
Weight (Metric) 18 g
Strap Attachment Type Elastic Straps
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.