Learn More
Sperian™ Honeywell™ FFP2 NRD Disposable Respirators
Product Code. 11846960
Change view
Click to view available options
Model:
FFP2D
Includes:
With Twin elasticstraps stapled outside the filtering area
With valve and full-face seal
Without valve For Single use
Unit Size:
Pack of 20
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Includes unitSize
11846960 FFP2D With Twin elasticstraps stapled outside the filtering area Pack of 20
11876900 FFP2D With valve and full-face seal Pack of 5
11811781 FFP2D Without valve For Single use Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11846960 Supplier Sperian™ Supplier No. 1005584
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Highly comfortable and ergonomically designed single-use respirators with no restriction with regard to use. Sperian™ Honeywell™ FFP2 NRD Respirators leverage the Willtech™ soft seal technology for an improved fit and exceptional comfort.
  • Rigid shell so shape is maintained even in moist and heavy loading environments
  • Hypoallergenic flange seal with absorption properties
  • Preformed and colored nose bridge to save time
  • Large internal volume to easily communicate and breathe
  • Twin elastic straps, ultra flexible and resistant, stapled outside of the filtering area
  • No latex, no silicone, no PVC
  • Protection and wearer acceptance guaranteed during periods of extended use
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Product Type Filtering Half Mask for Single Use
For Use With (Application) Textile, Mining, Construction, Iron and Steel industries, Car Body Preparation Industry, Woodworking.
Includes With Twin elasticstraps stapled outside the filtering area
Latex Free Yes
Material (Faceshield) PES knit, PU foam, PU film
Model FFP2D
Nose Piece Material Polypropylene-wrapped Galvanized wire
Quantity 20/Pk.
With Faceseal Yes
Weight (Metric) 10g