Learn More
3M™ FFP2 Unvalved Particulate Respirator Dispenser
Description
- Provides economic FFP2 (NPF 12) breathing protection against airborne particles and non-volatile liquids
- Flat-fold design makes the respirator easy to store when not in use
- Single-loop strap helps you make easy adjustments for a comfortable fit
- Diamond-style front panel helps the respirator retain shape even in hot or humid conditions
- Color-coded nose clip for easy identification: blue for FFP2
- Individually wrapped
- Diamond-shaped front panel allows the respirator to retain its shape even in hot or humid conditions
- Meets clogging requirements as per EN 149:2001+A1:2009, ensuring prolonged respirator life while maintaining ease of breathing
- Supplied in a handy dispenser box
- Noseclip is colored for easy identification: blue for FFP2
Specifications
Specifications
| Certifications/Compliance | CE Approved, EN 149: 2001 + A1: 2009 FFP2 NR D |
| Color | White |
| Product Type | Folded particle mask |