Learn More
FFP2 Disposable Respirator Face Mask
FFP2 Disposable respirator to protect against inhalation of droplets and particles. Available with head band and without exhalation valve.
Brand:  Medicom M52010(S)-WH
Product Code. 16629422
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Alert:
Compliance
This product complies with the essential requirements for health and safety as they are provided by the European Regulation (EU) 2016/425 for Personal Protective Equipment.
Description
Description
  • Tested and certified FFP2 disposable respirator
  • Without exhalation valve
  • Head bands enable a more secure, tighter and comfortable fit
  • Head bands make wearing glasses more comfortable and enable good compatibility with various eye protection products