missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian™ Honeywell™ Premium Disposable Respirators
Description
- Shielded exhalation valve for improved protection and breathing comfort
- Hypoallergenic seal with absorption properties
- Preformed and colored nose bridge to save time
- Large internal volume to easily communicate and breathe
- No latex, no silicone, no PVC
- Protection and wearer acceptance guaranteed during periods of extended use
Specifications
Specifications
| Disposable | Single-use |
| Product Type | Filtering Half Mask for Single Use, Flat fold |
| Size | Medium |
| With Exhalation Valve | Yes |
| Latex Free | Yes |
| Material (Faceshield) | PES knit, PU foam, PU film |
| Model | FFP1D |
| Nose Piece Material | Polypropylene-wrapped Galvanized wire |
| Quantity | 10/Pk. |
| Resistant To | Fluid |
| Show More |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?