missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FFAR3/GPR41 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03618-100UL
This item is not returnable.
View return policy
Description
FFAR3/GPR41 Polyclonal specifically detects FFAR3/GPR41 in Rat samples. It is validated for Western Blot.
Specifications
| FFAR3/GPR41 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| free fatty acid receptor 3, G protein-coupled receptor 41, GPR41FFA3R, G-protein coupled receptor 41 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 257-346 of human FFAR3/GPR41 (NP_005295.1). | |
| 100 μL | |
| GPCR | |
| 2865 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction