missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FFAR2 Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19952-1-AP-20UL
This item is not returnable.
View return policy
Description
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.Specifications
| FFAR2 | |
| Polyclonal | |
| Unconjugated | |
| FFAR2 | |
| FFA2R, FFAR2, free fatty acid receptor 2, G protein coupled receptor 43, GPR43 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100126285, 2867 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O15552 | |
| FFAR2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction