missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva FF170HP Membranes
Click to view available options
Dimensions (L x W):
210 x 297 mm
Unit Size:
Pack of 10
Description
FF High Performance (HP) membranes are part of the AE family, which are directly cast onto a plastic film.
- The FFHP membranes are a result of improved membrane casting procedures, which result in enhanced intra- and inter lot consistency and sharper lines.
- The surface is uniform without unincorporated nitrocellulose powder and the fine structure fiber distribution provides large internal surfaces for binding proteins
Nitrocellulose membranes are a key functional part of lateral-flow assays.
- The membrane must provide sufficient protein binding to enable the formation of a sharp and intense capture line, but at the same time the level of nonspecific background must be low for easy interpretation of results.
- Nitrocellulose membranes are available in a range of wicking rates and formulations.
- It has long been known that the wicking rate of a membrane has a significant impact on test sensitivity.
Specifications
Specifications
| Filter Type | Nitrocellulose |
| Material | Polyester |
| Sterility | Sterile |
| Flow Rate | 140-200 s/4cm |
| Quantity | 10/Pk. |
| Thickness (Metric) | 200 μm |
| Dimensions (L x W) | 210 x 297 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction