missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FEZ2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520456
This item is not returnable.
View return policy
Description
A suggested positive control is mouse brain tissue lysate. PA5-20456 can be used with blocking peptide PEP-0573.
Fasciculation and elongation protein zeta-2 (FEZ2) is a homolog to the mammalian FEZ1, itself an ortholog of the C. elegans UNC-76. In contrast to FEZ1, FEZ2 mRNA is widely expressed in mouse tissues. FEZ2 interacts with protein kinase C (PKC)-zeta substrate and induces neurite extension of PC12 cells when co-expressed with a constitutively active form of PKC-zeta, suggesting FEZ2 may play an important role in the morphological changes of various cells by associating with PKC-zeta in a tissue non-specific manner. FEZ2 can interact with FEZ1 through its c-terminal regions and especially its coiled-coil region. At least two isoforms of FEZ2 are known to exist.
Specifications
| FEZ2 | |
| Polyclonal | |
| Unconjugated | |
| FEZ2 | |
| 9030616F16; D17Ertd315e; fasciculation and elongation protein zeta 2; fasciculation and elongation protein zeta 2 (zygin II); fasciculation and elongation protein zeta-2; FEZ2; HUM3CL; pre-T/NK cell associated protein (3Cl); synaptotagmin interacting protein zyginII; Zrp; zygin 2; zygin II; Zygin-2; zygin-related protein types I/II | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 225020, 94269, 9637 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P97578, Q6TYB5, Q9UHY8 | |
| FEZ2 | |
| A 15 amino acid peptide from near the carboxy terminus of human FEZ2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction