Learn More
FEZ2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-81716H
Product Code. 18261147
  • £493.00 / 0.10mL
Description
FEZ2 Polyclonal antibody specifically detects FEZ2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FEZ2
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA
HRP
fasciculation and elongation protein zeta 2 (zygin II)
FEZ2 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human FEZ2. The immunogen is located within the last 50 amino acids of FEZ2. Amino Acid Squence: IPYEKKNGPPSVEDL
0.1 mL
Neuroscience
9637
Store at 4°C in the dark.