missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine Methyl Ester, TRC

Product Code. 30402654
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402654 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402654 Supplier Toronto Research Chemicals Supplier No. TRCF32251050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 2-[4-[(1RS)-1-Hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-butyl]phenyl]-2-methylpropanoate
CAS 154825-96-4
Molecular Formula C33 H41 N O4
Quantity 50 mg
Synonym Methyl 2-[4-[(1RS)-1-Hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-butyl]phenyl]-2-methylpropanoate
InChI Formula InChI=1S/C33H41NO4/c1-32(2,31(36)38-3)26-18-16-25(17-19-26)30(35)15-10-22-34-23-20-29(21-24-34)33(37,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29-30,35,37H,10,15,20-24H2,1-3H3
SMILES COC(=O)C(C)(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
IUPAC Name methyl 2-[4-[1-hydroxy-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butyl]phenyl]-2-methylpropanoate
Molecular Weight (g/mol) 515.6829
Formula Weight 515.3036
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.