missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine hydrochloride, MedChemExpress

Product Code. 16408628
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16408628

Brand: Medchem Express HYB0801A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fexofenadine hydrochloride (MDL-16455 hydrochloride), a H1R antagonist, is an anti-allergic agent used in seasonal allergic rhinitis and chronic idiopathic urticarial (person aged ≥16 years).

Fexofenadine hydrochloride (MDL-16455 hydrochloride), a H1R antagonist, is an anti-allergic agent used in seasonal allergic rhinitis and chronic idiopathic urticarial (person aged ≥16 years).

Specifications

Chemical Name or Material Fexofenadine hydrochloride
Physical Form Solid
CAS 153439-40-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C32H40ClNO4
For Use With (Application) COVID-19-immunoregulation
Synonym MDL-16455 hydrochlorideTerfenadine carboxylate hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (185.83 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)C(C)(C)C1=CC=C(C(O)CCCN2CCC(C(C3=CC=CC=C3)(O)C4=CC=CC=C4)CC2)C=C1.[H]Cl
Molecular Weight (g/mol) 538.12
Formula Weight 538.12
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.