missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine hydrochloride, MedChemExpress

Product Code. 16414497
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16414497 50 mg 50mg
16408628 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16414497 Supplier Medchem Express Supplier No. HYB0801A
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fexofenadine hydrochloride (MDL-16455 hydrochloride), a H1R antagonist, is an anti-allergic agent used in seasonal allergic rhinitis and chronic idiopathic urticarial (person aged ≥16 years).

Fexofenadine hydrochloride (MDL-16455 hydrochloride), a H1R antagonist, is an anti-allergic agent used in seasonal allergic rhinitis and chronic idiopathic urticarial (person aged ≥16 years).

Specifications

Chemical Name or Material Fexofenadine hydrochloride
Physical Form Solid
CAS 153439-40-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C32H40ClNO4
For Use With (Application) COVID-19-immunoregulation
Synonym MDL-16455 hydrochlorideTerfenadine carboxylate hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (185.83 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)C(C)(C)C1=CC=C(C(O)CCCN2CCC(C(C3=CC=CC=C3)(O)C4=CC=CC=C4)CC2)C=C1.[H]Cl
Molecular Weight (g/mol) 538.12
Formula Weight 538.12
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.