missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fexofenadine hydrochloride

Product Code. 16544304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16544304 250 mg 250mg
16534304 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16544304 Supplier Thermo Scientific Chemicals Supplier No. 464972500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 153439-40-8 | C32H40ClNO4 | 538.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153439-40-8
Molecular Formula C32H40ClNO4
Molecular Weight (g/mol) 538.13
MDL Number MFCD00865710
InChI Key RRJFVPUCXDGFJB-UHFFFAOYNA-N
IUPAC Name hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
SMILES [H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 538.12
Chemical Name or Material Fexofenadine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.