missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fexofenadine-d10 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Fexofenadine diphenyl-d10 hydrochloride |
| CAS | 1215821-44-5 |
| Molecular Formula | C32 2H10 H29 N O4 . Cl H |
| Quantity | 1 mg |
| Synonym | 4-[1-Hydroxy-4-[4-(hydroxydi(phenyl-d5)methyl)-1-piperidinyl]butyl]-α,α-di(methyl-d3)-benzeneacetic acid hydrochloride, Fexofenadine-d10 hydrochloride, Fexofenadine (diphenyl D10) hydrochloride |
| InChI Formula | InChI=1S/C32H39NO4.ClH/c1-31(2,30(35)36)25-17-15-24(16-18-25)29(34)14-9-21-33-22-19-28(20-23-33)32(37,26-10-5-3-6-11-26)27-12-7-4-8-13-27;/h3-8,10-13,15-18,28-29,34,37H,9,14,19-23H2,1-2H3,(H,35,36);1H/i3D,4D,5D,6D,7D,8D,10D,11D,12D,13D; |
| SMILES | Cl.[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(O)(C2CCN(CCCC(O)c3ccc(cc3)C(C)(C)C(=O)O)CC2)c4c([2H])c([2H])c([2H])c([2H])c4[2H] |
| IUPAC Name | 2-[4-[1-hydroxy-4-[4-[hydroxy-bis(2,3,4,5,6-pentadeuteriophenyl)methyl]piperidin-1-yl]butyl]phenyl]-2-methylpropanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 548.18 |
| Formula Weight | 547.3274 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?