missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FEV Recombinant Rabbit Monoclonal Antibody (HL2045)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550365
This item is not returnable.
View return policy
Description
FEV Recombinant Monoclonal Antibody for ICC/IF
This gene belongs to the ETS transcription factor family. ETS family members have a highly conserved 85-amino acid ETS domain that binds purine-rich DNA sequences. The alanine-rich C-terminus of this gene indicates that it may act as a transcription repressor. This gene is exclusively expressed in neurons of the central serotonin (5-HT) system, a system implicated in the pathogeny of such psychiatric diseases as depression, anxiety, and eating disorders. In some types of Ewing tumors, this gene is fused to the Ewing sarcoma (EWS) gene following chromosome translocations.
Specifications
| FEV | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q99581 | |
| FEV | |
| Full length human Pet1 recombinant protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| HL2045 | |
| Unconjugated | |
| FEV | |
| ETS domain transcription factor Pet-1; ETS-domain transcription factor Pet-1; Fev; FEV (ETS oncogene family); FEV (fifth Ewing variant); FEV transcription factor, ETS family member; FEV, ETS transcription factor; fifth Ewing variant protein; HSRNAFEV; mPet-1; PC12 ETS domain-containing transcription factor 1; PC12 ETS factor 1; Pet1; Pet-1; pheochromocytoma 12 ETS (E26 transformation-specific); protein FEV | |
| Rabbit | |
| Protein A | |
| RUO | |
| 54738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction