missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEV Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25058-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the ETS transcription factor family. ETS family members have a highly conserved 85-amino acid ETS domain that binds purine-rich DNA sequences. The alanine-rich C-terminus of this gene indicates that it may act as a transcription repressor. This gene is exclusively expressed in neurons of the central serotonin (5-HT) system, a system implicated in the pathogeny of such psychiatric diseases as depression, anxiety, and eating disorders. In some types of Ewing tumors, this gene is fused to the Ewing sarcoma (EWS) gene following chromosome translocations.Specifications
| FEV | |
| Polyclonal | |
| Unconjugated | |
| FEV | |
| FEV, FEV (ETS oncogene family), Fifth Ewing variant protein, HSRNAFEV, PC12 ETS factor 1, PET 1, PET1, Protein FEV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 260298, 54738 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8QZW2, Q99581 | |
| FEV | |
| FEV Fusion Protein Ag20647 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction