Learn More
FEV Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25058-1-AP-150UL
 View more versions of this product
Product Code. 16873595
  • £349.00 / 150µL
Description
This gene belongs to the ETS transcription factor family. ETS family members have a highly conserved 85-amino acid ETS domain that binds purine-rich DNA sequences. The alanine-rich C-terminus of this gene indicates that it may act as a transcription repressor. This gene is exclusively expressed in neurons of the central serotonin (5-HT) system, a system implicated in the pathogeny of such psychiatric diseases as depression, anxiety, and eating disorders. In some types of Ewing tumors, this gene is fused to the Ewing sarcoma (EWS) gene following chromosome translocations.
Specifications
Specifications
FEV
Polyclonal
Unconjugated
FEV
FEV, FEV (ETS oncogene family), Fifth Ewing variant protein, HSRNAFEV, PC12 ETS factor 1, PET 1, PET1, Protein FEV
Rabbit
Antigen Affinity Chromatography
RUO
260298, 54738
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8QZW2, Q99581
FEV
FEV Fusion Protein Ag20647
150 μL
Primary
Human, Mouse