missing translation for 'onlineSavingsMsg'
Learn More
Learn More
fetuin-B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18052-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.Specifications
| fetuin-B | |
| Polyclonal | |
| Unconjugated | |
| FETUB | |
| 16G2, FETUB, fetuin B, Fetuin like protein IRL685, Gugu, IRL685 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26998, 59083, 83928 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9QX79, Q9QXC1, Q9UGM5 | |
| FETUB | |
| fetuin-B Fusion Protein Ag12562 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction