missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fetuin B Monoclonal Antibody (K49014_4E12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555984
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: VNQKPTNLPK VEESQQKNTP PTDSPSKAGP RGSVQYLPDL DDKNSQEKGP QEAFPVHLDL TTNPQGETLD ISFLFLEPME
The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.
Specifications
| Fetuin B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9UGM5 | |
| FETUB | |
| Recombinant human FETUB (immunogen range is 271-350). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| K49014_4E12 | |
| Unconjugated | |
| FETUB | |
| 16G2; 2310011O17Rik; AI255764; D17980; Fet; FETUB; fetuin B; fetuin beta; fetuin phosphoprotein; fetuin-B; Fetuin-like protein IRL685; Gugu; IRL685; Pp63 | |
| Mouse | |
| Protein A | |
| RUO | |
| 26998 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction