missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin B Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98307F
This item is not returnable.
View return policy
Description
Fetuin B Polyclonal antibody specifically detects Fetuin B in Mouse samples. It is validated for ELISA.
Specifications
| Fetuin B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26998 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| FITC | |
| FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
| Produced in rabbits immunized with purified, recombinant Mouse Fetuin B (Q9QXC1-1; Met 1-Pro 388). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction