missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin B Antibody (110), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90643AF647
This item is not returnable.
View return policy
Description
Fetuin B Monoclonal antibody specifically detects Fetuin B in Mouse samples. It is validated for ELISA.
Specifications
| Fetuin B | |
| Monoclonal | |
| Alexa Fluor 647 | |
| FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Fetuin B (Q9QXC1-1; Met 1-Pro 388). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human FETUB | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 110 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 26998 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction