missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin A Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547157
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Approximately 5% cross-reactivity with recombinant human Fetuin A/AHSG is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Fetuin A Polyclonal antibody specifically detects Fetuin A in Human, Mouse samples. It is validated for Western Blot.Specifications
Fetuin A | |
Polyclonal | |
Unconjugated | |
AHSG | |
59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
11625, 197 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P02765, P29699 | |
AHSG | |
Mouse myeloma cell line NS0-derived recombinant mouse Fetuin A/AHSG Ala19-Ile345 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |