missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fetuin A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551593
This item is not returnable.
View return policy
Description
Immunogen sequence: LPPSTYVEFT VSGTDCVAKE ATEAAKCNLL AEKQYGFCKA TLSEKLGGAE VAVTCMVFQT QPVSSQPQPE GANEAVPTPV VDPDAPPSPP LGAPGLPPAG SPPDSHVLLA APPGHQLHRA HYDLRHTFMG VVSLGSPS Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 43%.
Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure.
Specifications
| Fetuin A | |
| Polyclonal | |
| Unconjugated | |
| AHSG | |
| 59 kDa bone sialic acid-containing protein; A2HS; Aa2-066; AHS; AHSG; alpha 2 HS-glycoprotein alpha 2 (fetuin); alpha 2-HS glycoprotein; alpha-2-HS-glycoprotein; Alpha-2-HS-glycoprotein chain A; Alpha-2-HS-glycoprotein chain B; Alpha-2-Z-globulin; asialofetuin; Ba-alpha-2-glycoprotein; BSP; Countertrypin; FETUA; fetuin; fetuin A; FetuinA; fetuin-A; Glycoprotein PP63; HSGA; phosphorylated N-glycoprotein pp63; pp63; PRO2743 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 197 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P02765 | |
| AHSG | |
| Recombinant protein corresponding to Human Fetuin A. Recombinant protein control fragment (Product #RP-100499). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction