missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin-A Mouse anti-Human, Clone: 1F6B9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Fetuin-A |
|---|---|
| Clone | 1F6B9 |
| Concentration | 0.96 mg/mL |
| Applications | Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16877655
|
Proteintech
66094-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16867655
|
Proteintech
66094-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human special AT-rich sequence-binding protein-2 (SATB2) is a nuclear matrix/scaffold-associated region DNA-binding protein. Like its homolog SATB1, SATB2 selectively binds double-stranded, special AT-rich DNA sequences, but is expressed primarily in a subset of postmitotic, differentiating neurons in the neocortex. Mice deficient in SATB exhibit craniofacial abnormalities and defects in osteoblast differentiation and function. SATB2 also interacts with and enhances the activity of Runx2 and ATF4, two transcription factors that regulate osteoblast differentiation, indicating that SATB2 acts as a molecular node in a transcriptional network regulating skeletal development and osteoblast differentiation. Recent experiments have shown that SATB2 interacts with histone deacetylase 1 and metastasis-associated protein 2, two proteins that are involved in chromatin remodeling, suggesting that SATB2 may also be involved in mediating epigenetic influences during cortical development.Specifications
| Fetuin-A | |
| 0.96 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2HS, AHS, AHSG, alpha 2 HS glycoprotein, Alpha 2 Z globulin, Ba alpha 2 glycoprotein, FETUA, Fetuin A, HSGA | |
| AHSG | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1F6B9 | |
| Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P02765 | |
| 197 | |
| Fetuin-A Fusion Protein Ag10073 | |
| Primary | |
| -20°C | |
| AHSG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title