missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin A/AHSG Antibody (SR2026), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22369-50ul
This item is not returnable.
View return policy
Description
Fetuin A/AHSG Monoclonal antibody specifically detects Fetuin A/AHSG in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Fetuin A/AHSG | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2026 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| A2HS, AHS, alpha-2-HS-glycoprotein, alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, FETUAba-alpha-2-glycoprotein, fetuin-A, HSGA | |
| A synthesized peptide derived from human Fetuin A/AHSG (Uniprot #: P02765) | |
| 50 μg | |
| Cancer | |
| 197 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction