missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Fetal Bovine Serum, ultra-low IgG, NZ origin

Click to view available options
Quantity:
500 mL
Unit Size:
Each
Description
Gibco fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera category: Specialty
- Origin: New Zealand
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
- IgG levels are less than 5 μg/mL and the BVD titer is low or not detectable.
Specifications
Specifications
| Content And Storage | Store at -5°C to-20°C. |
| Age | Fetal |
| Country of Origin | New Zealand |
| Format | Bottle |
| Quantity | 500 mL |
| Serum Treatment | Reduced IgG |
| Shipping Condition | Dry Ice |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction