missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, USDA-approved regions

Product Code. 11578836
Click to view available options
Quantity:
50 mL
1000 mL
10 x 50 mL
Format:
Bottle
One Shot
Unit Size:
1000mL
50mL
Pack of 10
This item is not returnable. View return policy

Product Code. 11578836

Brand: Gibco™ 10437036

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Important Update: We've recently introduced Value Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified USDA origin and Brazil (South American) origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Replacing with Fetal Bovine Serum, value
  • Sera application: for cell culture with robust cell lines, good value for basic research
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
  • Origin: countries meeting USDA importation requirements

Gibco serum delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Fetal
Country of Origin Mexico
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 1000 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Purity or Quality Grade Qualified
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.