missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, E.U.-approved, South America Origin

Product Code. 11550356
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11550356 500 mL 500mL
11543407 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11550356 Supplier Gibco™ Supplier No. 10500064

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C. results.

Sera Category: Standard
Origin: E.U. Approved (South American)
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C. results.
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Basic cell culture, specialty research, and specific assays
Certifications/Compliance EU-Approved
Content And Storage Store in freezer (–5 to –30°C)
Description Foetal Bovine Serum (Heat Inactivated), E.U. Approved (South American Origin), 500mL, Liquid, –5 to –30°C Storage Temperature, Cattle/Bovine Species, Virus and Mycoplasma Tested, Dry Ice
Country of Origin E.U. Approved (South American)
Form Liquid
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Source Cattle/Bovine
Grade Qualified
Tested For Virus, Mycoplasma
Formulation Liquid
For Use With (Application) Basic cell culture, specialty research, and specific assays
Heat Inactivated Heat-Inactivated
Purity or Quality Grade Qualified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.