missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Fetal Bovine Serum, US Characterized
Used for Cell culture, transfection.
Marca: Cytiva SH30071.02HI
170.73 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Characterized FBS
- Filtered through three sequential 100nm (0.1μm) pore sized rated filters
Especificaciones
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Characterized Fetal Bovine Serum, US Origin | |
Characterized | |
100 mL | |
Sequential Triple-filtered, 100 nm | |
Characterized | |
≤ 25 EU/mL | |
≤ 25 mg/dL | |
USDA Inspected |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
Fetal Bovine Serum | |
Sterile | |
Bovine Fetus | |
Heat Inactivated | |
Yes | |
No |